Surgical tapes, Durapore™
Leverandør: 3M MEDICAL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M1538-1
MMMEM1538-1EA 1350 DKK
MMMEM1538-1
Surgical tapes, Durapore™
Førstehjælpsudstyr Forbindinger
Durable, no-stretch silk-like cloth tape for critical tubing applications. Silk-like cloth, hypoallergenic tape, standard roll.
  • High strength, versatile, conformable
  • Adheres well to dry skin
  • Convenient bidirectional tear
  • Hypoallergenic and not made with natural rubber latex
  • Water resistant
Adhesion coupled with a strong backing with high load bearing capacity makes it ideal for securing critical tubings and devices and stabilising finger splints.