Powered air respirator air-flow adjustment covers, Versaflo™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S955
MMMAS955EA 152 DKK
MMMAS955
Powered air respirator air-flow adjustment covers, Versaflo™
Åndedrætsværn Åndedrætsbeskyttelsessystemer
The Versaflo™ Airflow Adjustment Cover S-955 is a spare airflow adjustment cover designed to replace worn or damaged parts in Versaflo™ S-Series Headtops.
  • Replacement airflow adjustment cover
  • Approved 3M replacement part
  • Extend product life by regularly checking for and replacing worn or damaged parts
This replacement part can extend the life of Versaflo™ S-Series Headtops. The airflow adjustment cover can be attached to the end of the breathing tube to allow the wearer to adjust the direction of the airflow in the hood to provide user comfort.