Powered and supplied air peel off visor covers and film for premium hoods, Versaflo™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S922
MMMAS922EA 381 DKK
MMMAS922
Powered and supplied air peel off visor covers and film for premium hoods, Versaflo™
Åndedrætsværn Åndedrætsbeskyttelsessystemer
The Versaflo™ Peel-Off Visor Cover for Premium Hoods is a transparent peel-off visor cover designed for use with Versaflo™ Series S-600, S-700, and S-800 premium head covers and hoods.
  • Protect visors from contaminants and scratches
  • For use with 3M™ Versaflo™ S-600, S-700 and S-800 premium headcovers and hoods
  • Self-adhesive cover can be easily applied and torn off
This peel-off visor cover can extend the service life of a visor by protecting it from contaminants and scratches during use. This self-adhesive cover can be easily applied and torn off.