Powered and supplied air peel off visor covers and films, Versaflo™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S920L S920S
MMMAS920LEA 381 DKK
MMMAS920L MMMAS920S
Powered and supplied air peel off visor covers and films, Versaflo™
Åndedrætsværn Åndedrætsbeskyttelsessystemer
Versaflo™ S Series Peel-Off Visor Covers are transparent, peel-off visor covers designed for use with Versaflo™ Series S-133, S-333, S-433, and S-533 head covers and hoods. This self-adhesive cover protects the visor from contaminants and scratches.
  • Protect visors from contaminants and scratches
  • Prolong the condition of visors
  • Self-adhesive cover can be easily applied and torn off
  • Available in large and small sizes
Prolong the condition of Versaflo™ S-133, S-333, S-433, and S-533 headcovers or hood visors with the Versaflo™ S Series Peel-Off VisorMore Product Information