Speedglas™ Inner Cover Plates
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H528015
MMMAH528015EA 379 DKK
MMMAH528015
Speedglas™ Inner Cover Plates
Svejseudstyr
Speedglas™ Inner protection plates are replaceable spare parts for 3M™ Speedglas™ welding helmets and are designed to protect the welding lens.
  • Helps protect the welding lens
  • Replace inner cover plate when it becomes deeply pitted or too dirty to clean
  • Regularly clean with a lint-free tissue or cloth
For maximum safety and comfort, regularly check equipment for worn or damaged parts.