Tape, grey, VHB™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
GPH-110GF
MMMAGPH-110GFEA 1280 DKK
MMMAGPH-110GF
Tape, grey, VHB™
Tape
3M™ VHB Tape GPH-110GF is a permanent bonding solution that can replace traditional mechanical fasteners and liquid adhesives in challenging, high temperature applications. Its acrylic foam core provides a good balance of strength and conformability. 3M™ VHB Tape GPH-110GF has excellent temperature resistance, ideal for powder coat or liquid paint processes which undergo a heat bake cycle.
  • High initial tack
  • Good balance of high temperature, and peel and shear performance
  • Foam enables stress relaxation and an easy application
  • Good sealing properties
  • For