Safety goggles, Fahrenheit™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FHEITAF TNWEAR FHEIT
MMMAFHEITAFEA 158 DKK
MMMAFHEITAF MMMATNWEAR MMMAFHEIT
Safety goggles, Fahrenheit™
Beskyttelsesbriller
3M™ Fahrenheit™ safety goggles feature a soft crystal frame and a wide support area around the face. They are designed with a specific channel on the side of the frame to accommodate the temple arms of prescription glasses.
  • Economical large profile safety goggle range, with enough space to fit over most prescription glassess
  • Polycarbonate lenses for impact resistance
  • 3M 1 BT lens marking
  • Indirect ventilation to protect against liquid droplets and large dust particles
  • Cylindrical curved lens design for optimal horizontal field of vision
Get comfortableMore Product Information