UHMW PE Film Tape, 5421
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5421101
MMMA5421101EA 5120 DKK
MMMA5421101
UHMW PE Film Tape, 5421
Tape
3M™ UHMW PE Film Tape 5421 protects surfaces from abrasion. This tape has a thickness 0,01 mm that offers excellent abrasion resistance, making it ideal for many tough impact wear or sliding applications. It creates a 'slip plane' effect to allow contact without wear. This is a thinner version of 3M™ UHMW-PE Film Tape 5423.
  • Excellent abrasion resistance
  • Achieve low coefficient of friction for ‘slip plane’ effect between surfaces
  • Reduce squeaks, rattles, and other noises that occur with movement
  • Use in applications where lubrication is not possible because