Boot covers, 440
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
440
MMMA440EA 3150 DKK
MMMA440
Boot covers, 440
Engangsbeklædning
These boot covers are made of combined SMS laminate and provide protection agaoinst non hazardous liquids and dust.
  • Anti-static treated
  • With ties for an excellent fit
  • Serged seams
Length: 406 mm.
Certificeringer: Cat. I.