Shoe covers, 402

Leverandør: 3M
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

402
MMMA402EA 1050 DKK
MMMA402
Shoe covers, 402
Engangsbeklædning
These disposable shoe covers made from now woven PP are designed to provide basic barrier protection to help protect footwear against light liquid splashes and hazardous dusts.

  • Elastic top and bottom for protection against contaminants and enhanced fit
  • Sole and upper constructed of the same material
  • Anti-static treated

Weight: 40 g/m²

Length: 250 - 380 mm
Height: 150 mm

Opening diameter: 1,5 - 15 cm
Bestil nu

Specifikationer

Environmentally Preferable
Stof PP
Antistatisk +
Køn
Outfit applikationer General Purpose,Medical,Laboratory,Industrial,Food
Weight 40 g/m²

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR