Microplate readers, Gemini™ XPS

Leverandør: Molecular Devices
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

XPS
MLDVXPSEA 0 DKK
MLDVXPS
Microplate readers, Gemini™ XPS
Mikropladelæsere
The Gemini™ XPS microplate spectrofluorometer provides a flexible environment to determine the optimal excitation and emission settings for most fluorescent intensity assays. The top reading Gemini™ XPS reader has dual monochromators that allow users to utilise new and novel dyes without purchasing expensive filter sets.

  • Read a variety of sample formats from 6- to 384-well microplates in endpoint, kinetic, spectral scan, and well scan modes
  • Eliminate the need for identifying, purchasing, and changing out filters for wavelength optimization with dual
More Product Information
Bestil nu

Specifikationer

Model Gemini™ XPS
Environmentally Preferable
Lyskilde Xenon flash lamp (1 joule/flash)
Detektor Photomultiplier (R-3896)
Wavelength range 250 - 850 nm (excitation), 360 - 850 nm (emission)
Valg af bølgelængde 1,0 nm increments
Wavelength accuracy ±2,0 nm
Wavelength bandwidth 9 nm
Incubation range Ambient +4 °C to 45 °C
Measurement time Reads a 96-well plate in <15 s, 384-well plate in <45 s
Software SoftMax® Pro
Temperature uniformity <1 °C at 37 °C set point
B×D×H 580×380×220 mm
Vægt 16 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR