Calcium assay kits, Fura-2 QBT™ Calcium kit
Leverandør: Molecular Devices
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
R8197 R8198
MLDVR8197EA 4240 DKK
MLDVR8197 MLDVR8198
Calcium assay kits, Fura-2 QBT™ Calcium kit
Assays Cellulær Assays
The Fura-2 QBT™ Calcium Kit eliminates the cause of data variability and reduces the number of steps compared to conventional wash protocols using Fura-2.
  • Largest Fura-2 signal window available
  • Eliminate wash artifacts and increase throughput with homogeneous assay
  • Minimise the impact of uneven dye loading and leakage on results
  • Interrogate low density, weakly or non adherent cells using no-wash protocol
  • Assay on FlexStation® 3 and FLIPR® Tetra Systems
  • Reference thousands of citations where Fura-2 has been used for cell lines and target
Leveringsinformation: For researchers concerned about assay artifacts, the Fura-2 QBT™ Calcium Kit utilises a simple, mix-and-read format that employs a proprietary masking technology with the industry-standard Fura-2 ratiometric calcium indicator to accurately measure calcium mobilisation.