Microplate reader, multi-mode, SpectraMax® Paradigm®
Leverandør: Molecular Devices
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PARADIGM
MLDVPARADIGMEA 0 DKK
MLDVPARADIGM
Microplate reader, multi-mode, SpectraMax® Paradigm®
Mikropladelæsere
The SpectraMax® Paradigm® reader can perform absorbance, fluorescence, time-resolved fluorescence (including HTRF), fluorescence polarisation, AlphaScreen®, AlphaLISA®, and luminescence measurements. Detection cartridge modularity allows users to configure the system to meet current needs, while providing the flexibility to address future applications.
  • Flexible design allows for single or dual excitation and emission, to address existing needs and future expansion
  • User upgradable in <2 minutes
  • High speed detection
  • Excellent sensitivity and performance