Millex® Syringe Filters, LCR Hydrophilic PTFE
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SLCR033NS SLCR033NB SLCR033NK SLLG033NS SLLG033NB SLLG033NK SLCRX13NK SLCRX13NL SLCRX13TL SLLGX13NK SLLGX13NL
MILFSLCR033NSEA 2490 DKK
MILFSLCR033NS MILFSLCR033NB MILFSLCR033NK MILFSLLG033NS MILFSLLG033NB MILFSLLG033NK MILFSLCRX13NK MILFSLCRX13NL MILFSLCRX13TL MILFSLLGX13NK MILFSLLGX13NL
Millex® Syringe Filters, LCR Hydrophilic PTFE
Filtre Sprøjtefiltre
Millex® Syringe filters are HPLC-certified for low levels of UV-absorbing extractables.
  • Lowest extractables, optimizing background levels of sensitive analyses like UHPLC and LC-MS
  • Filtration of protein-containing aqueous or organic solutions
  • Available in 0,20 μm and 0,45 μm pore sizes and three diameters to suit your application needs
  • Polypropylene housing material offers low levels of extractables and broad chemical compatibility
Applications are sample filtration prior to UHPLC, HPLC, LC-MS, and mass spec, Solvent filtration, Clarification ofMore Product Information