Samplicity® G2 Filtration System Tube Set Assembly
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SAMTUBING SAMVIALTR SAMWASTTR
MILFSAMTUBINGEA 500 DKK
MILFSAMTUBING MILFSAMVIALTR MILFSAMWASTTR
Samplicity® G2 Filtration System Tube Set Assembly
Filtre Topfiltre til flasker
Samplicity® G2 Filtration system enables vacuum driven filtration of up to 8 samples at once, directly into HPLC vials.
Filtration for HPLC sample prep without syringes. Sample is added to an adapter funnel on a standard Millex® 33 mm filter (Millex® Filters for the Samplicity® G2 System) or an integrated funnel and filter (Millex Samplicity® filter). Samplicity® G2 System has lower ergonomic impact than manual syringe filtration.
Applications: HPLC, Analytical sample preparation, Filtration of viscous or particulate laden samples for HPLC.