Millicell® µ-Migration Assay Kit
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MMA205
MILFMMA205EA 10330 DKK
MILFMMA205
Millicell® µ-Migration Assay Kit
Assays Cellulær Assays
This Kit overcomes the limitations of traditional multiwell migration assays. Its microfluidic, low-volume technology promotes a stable, diffusion-generated concentration gradient that is consistently linear and lasts for more than 48 hours.