Masterflex® L/S® Precision Pump Tubing, Peroxide-Cured Silicone, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX96400-13EA 814 DKK
MFLX96400-13 MFLX96400-14 MFLX96400-16 MFLX96400-17 MFLX96400-18 MFLX96400-25
Masterflex® L/S® Precision Pump Tubing, Peroxide-Cured Silicone, Avantor®
Slanger Peristaltisk pumpeslange
Excellent biocompatibility for a wide range of applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and