Tygon® 3350 Platinum-Cured Silicone Tubing

Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

95702-18
MFLX95702-00EA 1420 DKK
MFLX95702-00 MFLX95702-01 MFLX95702-02 MFLX95702-03 MFLX95702-04 MFLX95702-05 MFLX95702-06 MFLX95702-08 MFLX95702-09 MFLX95702-10 MFLX95702-13 MFLX95702-14 MFLX95702-16 MFLX95702-17 MFLX95702-19 MFLX95702-20 MFLX95702-18
Tygon® 3350 Platinum-Cured Silicone Tubing
Slanger Overførselsslange
Reduced particle entrapment with ultra-smooth inner surface.

  • Minimal extractables help maintain fluid integrity

Formulation: TYGON® 3350 silicone, platinum cured.
Applications: Pharmaceutical, cosmetic, industrial, and auto-analysis applications.
Characteristics: Ultra-smooth inner surface reduces particle entrapment and minimizes bacterial growth. Good for mild to medium concentration bases, salts, and alcohols; is odorless, tasteless, and non-toxic. Transparent.
Certification: Elastomer is FDA-compliant (21 CFR 177.2600); meets USP Class VIMore Product Information
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR