Masterflex® Ismatec® Reglo Independent Channel Control (ICC) Peristaltic Pumps, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX78001-58-DKEA 25740 DKK
MFLX78001-58-DK MFLX78001-60-DK MFLX78001-62-DK MFLX78001-68-DK MFLX78001-70-DK MFLX78001-72-DK MFLX78001-78-DK MFLX78001-80-DK MFLX78001-82-DK MFLX78001-58-EU MFLX78001-60-EU MFLX78001-62-EU MFLX78001-68-EU MFLX78001-70-EU MFLX78001-78-EU MFLX78001-80-EU MFLX78001-82-EU
Masterflex® Ismatec® Reglo Independent Channel Control (ICC) Peristaltic Pumps, Avantor®
Pumper Slangepumper
Independently control flow and direction of each channel via keypad or your PC.
  • Flow rate 0.001 to 43 ml/min per channel - flow rate depends on drive rpm, rollers, and tubing size
  • USB interface for quick connection to a PC
  • Flexibility of bidirectional flow in each channel
These multi-channel peristaltic pumps let you individually control the flow and direction of each channel from the intuitive keypad or via your computer—expanding your benchtop pumping capabilities with a single compact unit! Pump delivers continuous pumping or precision dispensingMore Product Information