Masterflex® EtherNet/IP™ Compatible Drives Powering Quattroflow™ Pumps
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX74000-49-DKEA 87140 DKK
MFLX74000-49-DK MFLX74000-51-DK MFLX74000-53-DK MFLX74000-59-DK MFLX74000-49-EU MFLX74000-51-EU MFLX74000-53-EU MFLX74000-59-EU
Masterflex® EtherNet/IP™ Compatible Drives Powering Quattroflow™ Pumps
Monitor transfer of delicate biological fluids with EtherNet/IP™ control capability.
  • Choose from readily available single-use or multiple-use Quattroflow™ pump head systems
  • Advanced network and communication models feature EtherNet/IP™
  • Diaphragms enable soft, gentle pumping through the strokes generated by the eccentric shaft
  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
  • Ideal for cross-flow systems, tangential flow filtration (TFF), chromatography, cultures, and more
The Quattroflow™More Product Information