Masterflex® Single-Use Ultrasonic Flow Sensor Controllers, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX33502-01EA 41410 DKK
MFLX33502-01
Masterflex® Single-Use Ultrasonic Flow Sensor Controllers, Avantor®
Flow switches
New Masterflex® Flow Controller automatically adjusts pump speed as tubing ages.
  • Compatible with all 33501-series of Masterflex® ultrasonic flow sensors
  • Proprietary technology
  • Simple operation
  • Data storage via USB interface
  • Compact footprint - only weighs two pounds
Using proprietary technology, the Masterflex® flow controller measures, controls, and logs fluid path information. It receives input from a flow sensor and then responds by adjusting the speed of your Masterflex pump to maintain a set flow rate or dispense volume. On the 4.3” LED touchMore Product Information