Masterflex® Normally Open Low-Flow Compact Switches, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
32778-32 32778-33 32778-34 32778-36
MFLX32778-30EA 1440 DKK
MFLX32778-30 MFLX32778-31 MFLX32778-35 MFLX32778-32 MFLX32778-33 MFLX32778-34 MFLX32778-36
Masterflex® Normally Open Low-Flow Compact Switches, Avantor®
Flow switches
Provides reliable operation in clean air and water applications.
  • Constructed of Noryl®, 316 stainless steel, and epoxy
These normally open SPST switches are specifically designed for reliable operation in clean air and water applications. Select from gas or liquid flow switches. Use a 50-µm filter to protect your switch. Switches include 18" (45.7 cm) L leads.
Leveringsinformation: Includes 18" (45.7 cm) L leads.