Hydrogen (H2) Gas Mass Flow Controllers with Metal bodies
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
32660-52 32660-58 32660-60 32660-66 32660-72 32661-58
MFLX32660-52EA 7910 DKK
MFLX32660-52 MFLX32660-58 MFLX32660-60 MFLX32660-66 MFLX32660-72 MFLX32661-58
Hydrogen (H2) Gas Mass Flow Controllers with Metal bodies
Flow switches
Precisely control flow rates with built-in valve.
  • Proven design uses metal body components for added durability
  • Accumulated flow can be logged using an optional totalizer
  • Includes NIST-traceable certificate
These controllers feature an advanced U-shaped sensor that ensures accurate and repeatable results. Gas flow measurements are unaffected by moderate temperature and pressure variations at the inlet. The meter also allows a four-point calibration across the flow range to improve meter linearity.
The flow rate set point can be established by either aMore Product Information