Masterflex® Industrial Flow Switches, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
32617-58 32617-60 32617-64 32617-66 32617-68
MFLX32617-58EA 1840 DKK
MFLX32617-58 MFLX32617-60 MFLX32617-64 MFLX32617-66 MFLX32617-68
Masterflex® Industrial Flow Switches, Avantor®
Flow switches
Designed for pump control in various applications and pipe sizes—continuously adjust while operating.
  • Easily interface with pumps for control pumping
These Masterflex® industrial flow switches are available in three model options. Choose from flow switch for use with corrosive liquids such as mild acids, flow switch for use in high-particle contaminated fluids, or flow switches that interface with chemical feed pumps.
Flow switch for use with corrosive liquids such as mild acids are designed for pump control in various applications and pipe sizes—comeMore Product Information