Kobold® Paddle Type Metal Flow Switches
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
32481-31 32481-32 32481-33 32481-34 32481-35 32481-36 32481-38 32481-41 32481-42
MFLX32481-31EA 1550 DKK
MFLX32481-31 MFLX32481-32 MFLX32481-33 MFLX32481-34 MFLX32481-35 MFLX32481-36 MFLX32481-38 MFLX32481-41 MFLX32481-42
Kobold® Paddle Type Metal Flow Switches
Flow switches
Adjust switch point to a maximum of 1450/3600 psi.
  • Switch point adjustable
  • Easy to install
  • Brass or Stainless Steel construction
  • Maximum pressure: 1450/3600 PSI
The device operates as follows: The flowing medium presses against the paddle for the flow switch. The paddle is fitted to one end of a balance arm which is in direct contact with a prestressed leaf spring. At the other end of the balance arm is a permanent magnet. This magnet actuates a reed contact located within a moveable housing outside the media. The reed contact switches on or offMore Product Information