Masterflex® Ismatec® Minicartridge Pump Heads for Masterflex® L/S® Drives, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX07623-00EA 7910 DKK
MFLX07623-00 MFLX07623-10 MFLX07623-30
Masterflex® Ismatec® Minicartridge Pump Heads for Masterflex® L/S® Drives, Avantor®
Pumper Pumpehoveder Peristaltiske pumpehoveder
Combine the accuracy of Ismatec® with the versatility of Masterflex® tubing pump systems.
  • Order optional minicartridges with adjustable occlusion to vary pressure each channel
  • Easily remove or insert minicartridges without disturbing other channels
  • Max pressure of 1.5 bar is achieved with pressure-lever cassettes only
  • Click-n-go cassettes offer a max pressure of 1.0 bar.
Choose from six, eight, or twelve roller heads. Less rollers offer greater flow rates, while more rollers offer a virtually pulseless flow. Heads accept up to four or eight Click’n’go™More Product Information