Masterflex® MasterSense™ MP30 Reusable Pressure Transducers, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX07526-82EA 10750 DKK
MFLX07526-82 MFLX07526-80
Masterflex® MasterSense™ MP30 Reusable Pressure Transducers, Avantor®
Pumper Slangepumper
High accuracy pressure transducers for plug and play connectivity with Masterflex® MasterSense™ pump systems.
  • Add critical pressure monitoring to your workflow
  • All-stainless welded body and wetted parts
  • Rugged design for shock protection and excellent long-term stability
  • CE certificated
  • Sanitary models are designed for clean-in-place and sterilize-in-place
asterSense™ MP30 pressure transducers utilize a thin-film-on-steel measurement principle to provide an accurate 0 to 30 psi output directly to MasterSense™ pump systems. Sensors include a six footMore Product Information