Masterflex® L/S® MasterSense™ Drives, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX07526-10-DKEA 37080 DKK
MFLX07526-10-DK MFLX07526-12-DK MFLX07526-20-DK MFLX07526-10-EU MFLX07526-12-EU MFLX07526-20-EU MFLX07526-22-DK MFLX07526-22-EU
Masterflex® L/S® MasterSense™ Drives, Avantor®
Pumper Pumpedrev Peristaltiske pumpedrev
Versatile pumps for accurate fluid delivery – with an intuitive touch-screen interface.
  • Powerful, space-efficient drives will accept all L/S pump head and tubing options
  • 5" (12,7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Pump lets you save and retrieve commonly used dispense programs for rapid set up
  • Over-the-air firmware updates let you take full advantage of improvements in drive features and functionality
  • Options for connectivity and monitoring include MasterflexLive, EtherNet/IP™, Profibus, or analog signal
  • Three-