Masterflex® L/S® Multichannel Cartridge Pump Head with Reduced Pulsation, 4-Channel, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX07519-20EA 5110 DKK
MFLX07519-20
Masterflex® L/S® Multichannel Cartridge Pump Head with Reduced Pulsation, 4-Channel, Avantor®
Pumper Pumpehoveder Peristaltiske pumpehoveder
Synchronous, low-pulsation flow from multiple channels.
  • Cartridges snap in and out for fast tubing changes—no need to remove pump head from the drive
  • Improved retainer design holds tubing securely in place
  • Flow rate 0.00006 to 85 ml/min with microbore 2-stop tubing—flow rate depends on drive rpm, cartridge, and tubing size
  • Accepts up to 4 large cartridges
  • Require drives with a minimum of 1⁄10 hp and a maximum of 250 rpm
  • Constructed with 8 Rulon® rollers, stainless steel rotor, and PSF ends
Each pump head system is composed of a heavy-duty pump head,More Product Information