Masterflex® L/S® Standard Pump Head Thrust Washers, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX07021-04EA 255 DKK
MFLX07021-04
Masterflex® L/S® Standard Pump Head Thrust Washers, Avantor®
Pumper Pumpehoveder Peristaltiske pumpehoveder
Get the most out of your Masterflex® L/S® Standard Pump Head with these replacement parts.
  • Suitable for use with Masterflex® L/S® Standard pump heads
Replacement parts to keep your pump head operating even longer. Thrust washers fit on either side of the rotor assembly of L/S standard pump heads.