Masterflex® L/S® High-Performance Precision Pump Tubing, Tygon® E-Lab, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX06509-15EA 1730 DKK
MFLX06509-15 MFLX06509-24 MFLX06509-35 MFLX06509-36
Masterflex® L/S® High-Performance Precision Pump Tubing, Tygon® E-Lab, Avantor®
Slanger Peristaltisk pumpeslange
Ideal for general transfer applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-