Rocker cell culture bags, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
91-200-92 91-200-78 91-200-79 91-200-80 91-200-81
MDTC91-200-92EA 3820 DKK
MDTC91-200-92 MDTC91-200-78 MDTC91-200-79 MDTC91-200-80 MDTC91-200-81
Rocker cell culture bags, Corning®
Poser Medieposer
Rocker cell culture bags are designed for use with commercial rocking motion bioreactor chambers, and feature irradiated ethylene vinyl acetate (EVA)/low-density polyethylene (ULDPE) 9101 film and stability bars on each side. These sterile, single-use cell culture bags are ideal for applications from basic research to large-scale biopharmaceutical manufacturing.
  • Provides non-invasive mixing of the cell culture/media with efficient gas transfer
  • Eliminates the risk of cross-contamination
  • Used for basic research to large scale cell culture applications