20X SSPE, liquid, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
46-021-CM
MDTC46-021-CMEA 2520 DKK
MDTC46-021-CM
20X SSPE, liquid, Corning®
Buffere Elektroforese buffere Hybridisation Buffers
20X Saline-Sodium-Phosphate-EDTA (SSPE) buffer has applications in nucleic acid hybridization and other molecular biology work.