Vand, Cell Culture Grade, tested to USP specifications for injection, sterile for sterile Water for Injection , Corning®

Leverandør: Corning
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Hydrogen oxide

25-055-LH 25-055-LG 25-055-LB 25-055-CVC 25-055-CM 25-055-CI 25-055-CV
MDTC25-055-LHEA 10320 DKK
MDTC25-055-LH MDTC25-055-LG MDTC25-055-LB MDTC25-055-CVC 25-055-CM MDTC25-055-CI 25-055-CV
Vand, Cell Culture Grade, tested to USP specifications for injection, sterile for sterile Water for Injection , Corning®
Vand

Corning® Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications. Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications.


  • Complete Certificate of Analysis available for each production lot
  • Produced under the highest industry standards to ensure superior results
  • Meets WFI Standards


Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).

Formel: H₂O
Molekylvægt: 18.02 g/mol
Kogepunkt: 100
Smeltepunkt: 0
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011332
CAS-nummer: 7732-18-5
Merck Index: 13,10098

Bestil nu

Specifikationer Testresultater

Mycoplasma Tested negative
Plating efficiency Pass
Oxidizable substances Solution remains faintly pink
Total organic carbon (before packaging) USP <643> < 500 ppb
Water conductivity (packaged water) USP <645> Meets USP Requirements
Water conductivity (before packaging) USP <645> Meets USP temperature/conductivity requirements
Endotoxin USP <85> ≤ 0.025 EU/mL
Sterility USP <71> Pass
Particulate matter USP <788> ≤ 25 particles/mL that are ≥ 10 μm in size
≤ 3 particles/mL that are ≥ 25 μm in size


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR