Trace elements C, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
25-023-CI
MDTC25-023-CIEA 506 DKK
MDTC25-023-CI
Trace elements C, Corning®
Mikrobiologiske medier Mikrobiologi Additiver og Supplementer
Trace Elements C Solution has the same composition as Cleveland's Trace Elements I, and contains the trace elements found in the serum-free medium MCDB301.