Avantor® ACE®, HPLC/UHPLC Columns, Extended Method Development Kits, 3 µm
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MDKE-3-2546UEA 14480 DKK
MDKE-3-2546U MDKE-3-2503U MDKE-3-2502U MDKE-3-1546U MDKE-3-1503U MDKE-3-1502U MDKE-3-1046U MDKE-3-1003U MDKE-3-1002U MDKE-3-0546U MDKE-3-0503U MDKE-3-0502U
Avantor® ACE®, HPLC/UHPLC Columns, Extended Method Development Kits, 3 µm
Kromatografikolonner
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (1,7 and 5 µm) and complementary phases
  • Analytical IDs in a