Avantor® ACE®, HPLC Columns, Bioanalytical 300 Å Method Development Kits, 5 µm
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MDKB-5-2546EA 12500 DKK
MDKB-5-2546 MDKB-5-2503 MDKB-5-2502 MDKB-5-1546 MDKB-5-1503 MDKB-5-1502 MDKB-5-1046 MDKB-5-1003 MDKB-5-1002 MDKB-5-0546 MDKB-5-0503 MDKB-5-0502
Avantor® ACE®, HPLC Columns, Bioanalytical 300 Å Method Development Kits, 5 µm
Kromatografikolonner
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (3 µm and 5 µm ) and complementary phases
  • Analytical IDs in a