Avantor® ACE®, HPLC/UHPLC Columns, Advanced Method Development Kits, 1,7 µm
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MDKA-17-1003UEA 11540 DKK
MDKA-17-1003U MDKA-17-1002U MDKA-17-0503U MDKA-17-0502U MDKA-17-0302U MDKA-17-D1002
Avantor® ACE®, HPLC/UHPLC Columns, Advanced Method Development Kits, 1,7 µm
Kromatografikolonner
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (1,7 and 5 µm) and complementary phases
  • Analytical IDs in a