VWR® Protease Inhibitor Cocktail, Plant
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M307-1MLEA 892 DKK
M307-1ML
VWR® Protease Inhibitor Cocktail, Plant
Celledyrkningssupplementer og additiver
Contains a mix of protease inhibitors: AEBSF, E-64, Bestatin, Pepstatin, Leupeptin and 1, 10-Phenanthroline. Each vial can be reconstituted in 1 ml dimethyl sulfoxide to form a 100X solution.