Magnesiumacetat tetrahydrat 99, reagent

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate

M0631-1KG
M0631-1KGEA 1480 DKK
M0631-1KG
Magnesiumacetat tetrahydrat 99, reagent
Magnesiumacetat tetrahydrat

  • ≥95% for general laboratory use.

Formel: (H₃CCOO)₂Mg·4H₂O
Molekylvægt: 214.46 g/mol
Smeltepunkt: 68 °C
Massefylde: 1.45 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149214
CAS-nummer: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676

Bestil nu

Specifikationer Testresultater

Assay 99 - 102 %
Insoluble in acetic acid Max. 0.005 %
Form Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR