Metylorange solution_SC 0,1%

Leverandør: Lovibond
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine

419436
LOVI419436EA 180 DKK
LOVI419436
Metylorange solution_SC 0,1%
Metylorange
Formel: C₁₄H₁₄N₃NaO₃S
Molekylvægt: 327.34 g/mol
Kogepunkt: 90
Flammepunkt: 38
MDL Number: MFCD00007502
CAS-nummer: 547-58-0
Merck Index: 13,06127

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR