Perfection and Performance laboratory refrigerators with plastic inner liner, SRFvh and SRFvg
Leverandør: Liebherr
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SRFVG4001 SRFVG4011 SRFVG5511 SRFVH5501
471-1337EA 18749 DKK
471-1337 LIEBSRFVG4011 471-1338 LIEBSRFVH5501
Perfection and Performance laboratory refrigerators with plastic inner liner, SRFvh and SRFvg
Køleskabe
Glass/steel door refrigerators, Performance series with ceiling lighting and perfection series with vertical LED lighting provides optimal illumination of the interior over the entire height of the appliance.
  • Alarm relay, alarm test function and custom alarm settings possible
  • +2 °C Safety Device function, Min./max. temperature log
  • Self-closing door at <90° and mechanical lock provided
  • Performance series with super-efficient and perfection series with ultra-efficient energy consumption
  • Internal liner material made of commercial grade moulded polystyrol