Refrigerators, BioFresh, upright
Leverandør: Liebherr
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
KB3660 KB4260 KBES3660 KBES4260 KB3160 KBES3160 B2756 KB4210
LIEBKB3660EA 0 DKK
LIEBKB3660 LIEBKB4260 LIEBKBES3660 LIEBKBES4260 LIEBKB3160 LIEBKBES3160 LIEBB2756 LIEBKB4210
Refrigerators, BioFresh, upright
Køleskabe
These are ideal for fresh food at home or house-hold type environment. This includes hotels, homes, motels, lodging, catering and similiar services.
  • The LED ceiling light creates a high-class lighting ambience within the refrigerator
  • GlassLine storage shelves
  • The door seal is designed to match the appliance door or housing colour