Performance Professional Refrigerators with Dynamic cooling and Plastic Inner Liner, FRFvg
Leverandør: Liebherr
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FRFVG5501 FRFVG4001 FRFVG6501
LIEBFRFVG5501EA 14359 DKK
LIEBFRFVG5501 LIEBFRFVG4001 LIEBFRFVG6501
Performance Professional Refrigerators with Dynamic cooling and Plastic Inner Liner, FRFvg
Køleskabe
Refrigerators designed for professional use. Reliably store non temperature-sensitive items even at ambient temperatures up to +40 °C with a constant cooling power. They are enormously robust, long-lasting, easy to clean and particularly sustainable, not only thanks to natural refrigerant. At the same time, they fulfil all the valid directives, standards and norms, including in terms of hygiene, HACCP concept and GN containers. The efficient cooling system not only ensures low energy and overall operating cost, also safe storage of items not requiring validation of temperature stability andMore Product Information
Bestil nu
Specifikationer
Model FRFvg 5501 FRFvg 4001 FRFvg 6501
Environmentally Preferable
Display Monochrome display, touch buttons, external digital Monochrome display, touch buttons, external digital Monochrome display, touch buttons, external digital
Defrost Automatic Automatic Automatic
Alarm Visual and audible Visual and audible Visual and audible
Shelves/drawers 4 adjustable, wire shelves plastic-coated 5 adjustable, wire shelves plastic-coated 5 adjustable, wire shelves plastic-coated
Dørtype Steel, self closing Steel, self closing Steel, self closing
Dørlås Mechanical Mechanical Mechanical
Isolering 45/57 mm 45/57 mm 45/57 mm
Kølemiddel R 600a R 600a R 600a
Sound level 49 dB(A) 48 dB(A) 49 dB(A)
Energy class C C C
Elektriske specifikationer 220 - 240 V~, 2,0 A, 50 Hz 220 - 240 V~, 2,0 A, 50 Hz 220 - 240 V~, 2,0 A, 50 Hz
Vægt 87 (gross)/78 (net) kg 70 (gross)/65 (net) kg 93 (gross)/84 (net) kg