Combined fridge freezers, Premium BioFresh, CBNPes series
Leverandør: Liebherr
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CBNPES3956 CBNPES3756 CBNPES3967 CBNPES3976
LIEBCBNPES3956EA 0 DKK
LIEBCBNPES3956 LIEBCBNPES3756 LIEBCBNPES3967 LIEBCBNPES3976
Combined fridge freezers, Premium BioFresh, CBNPes series
Køleskabe
These combined fridge freezers with biofresh and NoFrost offer more interior capacity for food storage, economy and energy efficiency.
  • SwingLine design
  • Stainless steel slimline handles with integrated opening mechanism
  • Versatile installation due to front ventilation
  • Transport handles at back
  • Height-adjustable feet at front, transport castors at back
  • Reversible door hinging
These freezers feature stainless steel door with SmartSteel and aluminium door handle with integrated opening mechanism. The NoFrost system provides rapid, fan-assisted, frost-freeMore Product Information