Erlenmeyer-kolber, bred åbning, med krave
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
07025022 07025028 07025037 07025046 07025049 07025052 07025058 07025070 07025073
LENZ07025022EA 813 DKK
LENZ07025022 LENZ07025028 LENZ07025037 LENZ07025046 LENZ07025049 LENZ07025052 LENZ07025058 LENZ07025070 LENZ07025073
Erlenmeyer-kolber, bred åbning, med krave
Kolber Erlenmeyer-kolber
DURAN®, borosilicate glass 3,3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area.