Reaction vessels, cylindrical
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
06000479 06000482 06000494 06000270 06001270 06000137 06000149 06000158 06000249 06000258 06000473 06000476 06000485 06000488 06000588 06000594 06001137 06001149 06001158 06001170 06001249 06001258 06001276 06001279 06001473 06001476 06001479 06001482 06001485 06001488 06001494 06001585 06001588 06001594 06000273 06001273 06000279
LENZ06000479EA 5140 DKK
LENZ06000479 LENZ06000482 LENZ06000494 LENZ06000270 LENZ06001270 LENZ06000137 LENZ06000149 LENZ06000158 LENZ06000249 LENZ06000258 LENZ06000473 LENZ06000476 LENZ06000485 LENZ06000488 LENZ06000588 LENZ06000594 LENZ06001137 LENZ06001149 LENZ06001158 LENZ06001170 LENZ06001249 201-1160 LENZ06001276 LENZ06001279 LENZ06001473 LENZ06001476 LENZ06001479 LENZ06001482 LENZ06001485 LENZ06001488 LENZ06001494 LENZ06001585 LENZ06001588 LENZ06001594 LENZ06000273 LENZ06001273 LENZ06000279
Reaction vessels, cylindrical
Reaktionsbeholdere Reaktionsbeholdere, cylinderformede
DURAN®, borosilicate glass, clear, with round bottom.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With easily readable graduation and laboratory flat flange (LF) with groove.