Powder filtered systems, XPert™
Leverandør: LABCONCO
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3961622
LBCP3961622EA 0 DKK
LBCP3961622
Powder filtered systems, XPert™
Stinkskabe og kabinetter Ekstraktionsskabe
XPert™ Bulk Powder Filtered System is designed to provide user protection from hazardous airborne particulates and fumes generated during powder transfer operations. Patented features include an ergonomic air foil with Clean-Sweep openings to enhance airflow and a zone-perforated rear baffle that creates horizontal laminar airflow to maximize containment. The system includes a built-in blower, 99,99% efficient HEPA filter, true bag-in/bag-out filter disposal system, and pre-wired fluorescent light and switches for blower and light.
  • With tempered safety glass front sash and sides ;