Biological safety cabinet, class II, type A2, Purifier® Logic®+
Leverandør: LABCONCO
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
302481030 302481150 302611040 302681150 302411030 302581041
LBCP302481030EA 0 DKK
LBCP302481030 LBCP302481150 LBCP302611040 LBCP302681150 LBCP302411030 LBCP302581041
Biological safety cabinet, class II, type A2, Purifier® Logic®+
Stinkskabe og kabinetter Biologiske sikkerhedskabinetter
Purifier Logic+ Class II, Type A2 Biosafety Cabinets provide personnel, product and environmental protection from hazardous particulates such as agents that require Biosafety Level 1, 2 or 3 containment. Other appropriate applications include work with antineoplastic drugs, genetic material, asbestos and additional substances that generate hazardous airborne particulates. These cabinets are designed to work with antineoplastic drugs, genetic material, asbestos and additional substances that generate hazardous airborne particulates. When canopy connected, the biosafety cabinets may be usedMore Product Information