Biosafety cabinets, Class II A2, Purifier®
Leverandør: LABCONCO
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
302411131 302381131 302481131 302611131 302581131 302611031 302380131
LBCP302411131EA 0 DKK
LBCP302411131 LBCP302381131 LBCP302481131 LBCP302611131 LBCP302581131 LBCP302611031 LBCP302380131
Biosafety cabinets, Class II A2, Purifier®
Stinkskabe og kabinetter Biologiske sikkerhedskabinetter
Purifier Logic+ Class II Type A2 Biosafety Cabinets are suitable for work with agents that require Biosafety Level 1, 2 or 3 containment.
  • Electronically Commutated Motor (ECM)
  • Constant Airflow Profile™ (CAP) Technology airflow monitoring system that communicates with the ECM to deliver the precise volume of air as required and automatically adjust as filters load without relying on airflow sensors
  • Line of sight LCD information center with Filter Life Remaining bar graph, status line for alarm conditions and alerts to warn when filter life diminishes to 20%